4-Aminophenylboronsäure Hydrochlorid 97%

Lieferant: Thermo Fisher Scientific
Achtung

H27479.MC H27479.ME
H27479.MCEA 74.1 EUR
H27479.MC H27479.ME
4-Aminophenylboronsäure Hydrochlorid 97%
4-Aminophenylboronsäure Hydrochlorid

Purity: 97% CAS No.: 80460-73-7 Molecular Formula: C6H8BNO2.HCl Formula Weight: 173.41 Melting Point: 125-132° MDL No.: MFCD03001333

Formel: C₆H₉BClNO₂
Schmelzpunkt: 125…132 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD03001333
CAS-Nummer: 80460-73-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR