DL-Methioninsulfoxid ≥98%

Lieferant: Thermo Fisher Scientific

Synonyme: 2-Amino-4-(methylsulphinyl)butanoic acid

A18081.06 A18081.14
A18081.06EA 57.1 EUR
A18081.06 A18081.14
DL-Methioninsulfoxid ≥98%
DL-Methioninsulfoxid

Purity: 98+% CAS No.: 62697-73-8 Molecular Formula: C5H11NO3S Formula Weight: 165.21 Melting Point: ca 230° dec. MDL No.: MFCD00002620

Formel: C₅H₁₁NO₃S
Schmelzpunkt: ∼ 230 °C (dec.)
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00002620
CAS-Nummer: 62697-73-8
EINECS: 263-700-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR