DL-Methioninsulfon ≥98%
Lieferant: Thermo Fisher Scientific
B25094.03
B25094.06
B25094.03EA
40.9
EUR
B25094.03
B25094.06
DL-Methioninsulfon ≥98%
DL-Methioninsulfon
Purity: 98%
CAS No.: 820-10-0
Molecular Formula: C5H11NO4S
Formula Weight: 181.21
MDL No.: MFCD00025079
Formel:
C₅H₁₁NO₄S Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00025079 CAS-Nummer: 820-10-0 EINECS: 212-466-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...