Cholesterololeylcarbonat 99+%

Lieferant: Thermo Fisher Scientific

Synonyme: (3S,8S,9S,10R,13R,14S,17R)-10,13-Dimethyl-17-((R)-6-methylheptan-2-yl)-2,3,4,7,8,9,10,11,12,13,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-3-yl (Z)-octadec-9-en-1-yl carbonate

165850050 165850250
ACRO165850050EA 36.7 EUR
ACRO165850050 ACRO165850250
Cholesterololeylcarbonat 99+%
Cholesterololeylcarbonat
Formel: C₄₆H₈₀O₃
Schmelzpunkt: 20 °C
Flammpunkt: >112 °C
MDL Number: MFCD00003634
CAS-Nummer: 17110-51-9
EINECS: 241-179-7

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow Waxy crystalline mass
Infrared spectrum Conforms
TLC ≥99.0 %
Specific optical rotation -22° to -24° (27°C, 589 nm) (c=2, CHCl3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR