Tetrathiafulvalen 99+%
Lieferant: Thermo Fisher Scientific
Synonyme:
2,2'-Bi(1,3-dithiolylidene), TTF
179410010
179412500
ACRO179410010EA
190
EUR
ACRO179410010
ACRO179412500
Tetrathiafulvalen 99+%
Tetrathiafulvalen
Formel:
C₆H₄S₄ Schmelzpunkt: 117…120 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00005492 CAS-Nummer: 31366-25-3 EINECS: 250-593-7 |
Spezifikation Testergebnisse
Appearance | Orange to brown Crystalline powder or crystals |
Infrared spectrum | Conforms |
Melting point | 117 to 120 °C |
GC | ≥99.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...