Methylmagnesiumchlorid ∼22% (w/w) 3 mol/l in THF, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ200
189901-100ML
189901-800ML
189901-8L
SIAL189901-100MLEA
64.2
EUR
SIAL189901-100ML
SIAL189901-800ML
SIAL189901-8L
Methylmagnesiumchlorid ∼22% (w/w) 3 mol/l in THF, Sigma-Aldrich®
Methylmagnesium chloride
Formel:
CH₃ClMg |
MDL Number:
MFCD00000468 CAS-Nummer: 676-58-4 EINECS: 211-629-7 UN-Nummer: 3399 ADR: 4.3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...