4'-Methylbiphenyl-4-carbonsäure 96%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
4'-Methyl-[1,1'-biphenyl]-4-carboxylic acid, 4'-Methyl-4-biphenylcarboxylic acid
H34459.03
H34459.06
H34459.03EA
66.3
EUR
H34459.03
H34459.06
4'-Methylbiphenyl-4-carbonsäure 96%
4'-Methylbiphenyl-4-carbonsäure
Formel:
C₁₄H₁₂O₂ Schmelzpunkt: 257…258 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00448778 CAS-Nummer: 720-73-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...