2'-Nitroacetanilid
Lieferant: Thermo Scientific
Synonyme:
N-(2-Nitrophenyl)acetamide
TL00610.1
TL00610.10
TL00610.50
MAYBTL00610.1EA
0
EUR
MAYBTL00610.1
MAYBTL00610.10
MAYBTL00610.50
2'-Nitroacetanilid
2'-Nitroacetanilid
Formel:
C₈H₈N₂O₃ Molekulargewicht: 180.16 g/mol Dichte: 1 g/cm³ (20 °C) |
MDL Number:
MFCD00016991 CAS-Nummer: 552-32-9 EINECS: 209-009-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...