Rhodium(III)oxid, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ100
204226-1G
204226-500MG
SIAL204226-1GEA
510
EUR
SIAL204226-1G
SIAL204226-500MG
Rhodium(III)oxid, Sigma-Aldrich®
Rhodium(III) oxide
Formel:
O₃Rh₂ |
MDL Number:
MFCD00011205 CAS-Nummer: 12036-35-0 EINECS: 234-846-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...