6-Brom-2,3-difluortoluol 95%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
1-Bromo-3,4-difluoro-2-methylbenzene
H32941.03
H32941.06
H32941.03EA
88.3
EUR
H32941.03
H32941.06
6-Brom-2,3-difluortoluol 95%
6-Brom-2,3-difluortoluol
Purity: 97%
CAS No.: 847502-81-2
Molecular Formula: C7H5BrF2
Formula Weight: 207.02
Refractive Index: 1.5105
Formel:
C₇H₅BrF₂ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD07777161 CAS-Nummer: 847502-81-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...