2,6-Bis(trifluormethyl)phenylboronsäure ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
2,6-Bis(trifluoromethyl)benzeneboronic acid
L19952.03
L19952.06
L19952.MD
L19952.03EA
89.6
EUR
L19952.03
L19952.06
L19952.MD
2,6-Bis(trifluormethyl)phenylboronsäure ≥97%
2,6-Bis(trifluormethyl)phenylboronsäure
Purity: 97%
CAS No.: 681812-07-7
Molecular Formula: C8H5BF6O2
Formula Weight: 257.93
Melting Point: ca 150°
MDL No.: MFCD04039321
Formel:
C₈H₅BF₆O₂ Schmelzpunkt: ∼ 150 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD04039321 CAS-Nummer: 681812-07-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...