4-tert-Butyl-2-methylthiazol

Lieferant: Thermo Scientific

CD09457.1 CD09457.50
MAYBCD09457.1EA 0 EUR
MAYBCD09457.1 MAYBCD09457.50
4-tert-Butyl-2-methylthiazol
4-tert-Butyl-2-methylthiazol
Formel: C₈H₁₃NS
MDL Number: MFCD00051643
CAS-Nummer: 15679-11-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR