4-(4-Bromphenyl)-2-methylpyrimidin

Lieferant: Thermo Scientific

KM02325.250 KM02325.1
MAYBKM02325.250EA 0 EUR
MAYBKM02325.250 MAYBKM02325.1
4-(4-Bromphenyl)-2-methylpyrimidin
4-(4-Bromophenyl)-2-methylpyrimidine
Formel: C₁₁H₉BrN₂
MDL Number: MFCD02089754
CAS-Nummer: 499785-50-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR