2-(Methylthio)thiophen ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 2-(Mehtylthio)thiophene

A11763.03 A11763.06
A11763.03EA 49.5 EUR
A11763.03 A11763.06
2-(Methylthio)thiophen ≥97%
2-(Methylthio)thiophene

Purity: 97% CAS No.: 5780-36-9 Molecular Formula: C5H6S2 Formula Weight: 130.23 Refractive Index: 1.5960 Odour: Stench Boiling Point: 82-84°/15mm Beilstein Reference: 106272 UN No.: 3334 MDL No.: MFCD00052382

Formel: C₅H₆S₂
Siedepunkt: 82…84 °C (15 mmHg)
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00052382
CAS-Nummer: 5780-36-9
ADR: 9,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR