3-Brom-5-chlor-2-thiophensulfonylchlorid

Lieferant: Thermo Scientific

SEW02653.250 SEW02653.1
MAYBSEW02653.250EA 0 EUR
MAYBSEW02653.250 MAYBSEW02653.1
3-Brom-5-chlor-2-thiophensulfonylchlorid
3-Brom-5-chlor-2-thiophensulfonylchlorid
Formel: C₄HBrCl₂O₂S₂
MDL Number: MFCD00052095
CAS-Nummer: 175205-72-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR