3,5-Bis(trifluormethyl)benzhydrazid ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
A13492.03
A13492.06
A13492.14
A13492.03EA
35.1
EUR
A13492.03
A13492.06
A13492.14
3,5-Bis(trifluormethyl)benzhydrazid ≥97%
3,5-Bis(trifluormethyl)benzhydrazid
Purity: 97%
CAS No.: 26107-82-4
Molecular Formula: C9H6F6N2O
Formula Weight: 272.15
Melting Point: 132-134°
Beilstein Reference: 2596448
Formel:
C₉H₆F₆N₂O Schmelzpunkt: 132…134 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00051848 CAS-Nummer: 26107-82-4 EINECS: 247-462-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...