4,4'-Anisoin ≥97%
Lieferant: Thermo Fisher Scientific
B21559.14
B21559.36
B21559.14EA
35.3
EUR
B21559.14
B21559.36
4,4'-Anisoin ≥97%
4,4'-Anisoin
Purity: 97%
CAS No.: 119-52-8
Molecular Formula: C16H16O4
Formula Weight: 272.30
Melting Point: 110-114°
Beilstein Reference: 3211892
Formel:
C₁₆H₁₆O₄ Schmelzpunkt: 110…114 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00008411 CAS-Nummer: 119-52-8 EINECS: 204-330-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...