4-(Hydroxymethyl)phenylboronsäurepinakolester

Lieferant: Thermo Fisher Scientific

Synonyme: 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl alcohol, 2-[4-(Hydroxymethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-(Hydroxymethyl)benzeneboronic acid pinacol ester, (4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)methanol, 4-(Hydroxymethyl)phenylboronic acid Pinacol ester

H51123.03 H51123.06 H51123.14
H51123.03EA 56.5 EUR
H51123.03 H51123.06 H51123.14
4-(Hydroxymethyl)phenylboronsäurepinakolester
4-(Hydroxymethyl)phenylboronsäurepinakolester

CAS No.: 302348-51-2 Molecular Formula: C13H19BO3 Formula Weight: 234.10 MDL No.: MFCD09837617

Formel: C₁₃H₁₉BO₃
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD09837617
CAS-Nummer: 302348-51-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR