4-(Methylsulfanyl)benzolthiol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

662496-10G 662496-1G
SIAL662496-10GEA 315 EUR
SIAL662496-10G SIAL662496-1G
4-(Methylsulfanyl)benzolthiol, Sigma-Aldrich®
4-(Methylthio)thiophenol
Formel: C₇H₈S₂
MDL Number: MFCD00082770
CAS-Nummer: 1122-97-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR