Kaliumvinyltrifluoroborat, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Potassium trifluoro(vinyl)borate
M-Clarity™ quality level = MQ100
655228-1G
655228-25G
655228-5G
SIAL655228-1GEA
45.3
EUR
SIAL655228-1G
SIAL655228-25G
SIAL655228-5G
Kaliumvinyltrifluoroborat, Sigma-Aldrich®
Kaliumvinyltrifluoroborat
Formel:
C₂H₃BF₃K |
MDL Number:
MFCD02093335 CAS-Nummer: 13682-77-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...