DL-Penicillamin 97+%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
2-Amino-3-mercapto-3-methylbutanoic acid
B24710.03
B24710.06
B24710.03EA
35.9
EUR
B24710.03
B24710.06
DL-Penicillamin 97+%
DL-Penicillamin
Purity: 97+%
CAS No.: 52-66-4
Molecular Formula: C5H11NO2S
Formula Weight: 149.21
Storage Temperature: 2°C to 8°C
Odour: Unpleasant
Beilstein Reference: 2039817
Merck Reference: 14,7088
MDL No.: MFCD00004856
Formel:
C₅H₁₁NO₂S Schmelzpunkt: ∼ 212 °C (dec.) |
MDL Number:
MFCD00004856 CAS-Nummer: 52-66-4 EINECS: 200-147-2 Merck Index: 14,7088 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...