1,2-Epoxycycloheptan, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

672440-1G 672440-5G
SIAL672440-1GEA 166 EUR
SIAL672440-1G SIAL672440-5G
1,2-Epoxycycloheptan, Sigma-Aldrich®
1,2-Epoxycycloheptane
Formel: C₇H₁₂O
MDL Number: MFCD00022357
CAS-Nummer: 286-45-3
EINECS: 206-009-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR