Tricain-Methansulfonat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: TS 222, 3-Aminobenzoic acid Ethyl Ester Methanesulphonate, Tricaine, Ethyl 3-aminobenzoate methanesulphonate, Ethyl 3-aminobenzoate, methanesulphonic acid salt, Bio-Calm™, Ethyl-m-Aminobenzoate, Methanesulphonate salt, MS-222

M-Clarity™ quality level = MQ100

E10521-10G E10521-50G
SIALE10521-10GEA 59.9 EUR
SIALE10521-10G SIALE10521-50G
Tricain-Methansulfonat, Sigma-Aldrich®
Tricain-Methansulfonat
Formel: C₁₀H₁₅NO₅S
MDL Number: MFCD00013176
CAS-Nummer: 886-86-2
EINECS: 212-956-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR