Samarium(II)iodid 0.1 mol/l in THF stabilisiert, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Samarium (II) iodide
Stabilisierung:
Stabilised with Sm
M-Clarity™ quality level = MQ200
347116-100ML
347116-25ML
SIAL347116-100MLEA
176
EUR
SIAL347116-100ML
SIAL347116-25ML
Samarium(II)iodid 0.1 mol/l in THF stabilisiert, Sigma-Aldrich®
Samarium diiodide
Formel:
SmI₂ |
MDL Number:
MFCD00058873 CAS-Nummer: 32248-43-4 UN-Nummer: 3271 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...