Fmoc-D-phenylalanin
Lieferant: Novabiochem (Merck Biosciences)
Synonyme:
Fmoc-D-Phe-OH, N-Fmoc-D-phenylalanine
8.52148.0025
8.52148.0005
04-13-1030-25EA
306
EUR
04-13-1030-25
04-13-1030-5
Fmoc-D-phenylalanin
Fmoc-D-phenylalanin
Formel:
C₂₄H₂₁NO₄ Molekulargewicht: 387.4 g/mol |
MDL Number:
MFCD00062955 CAS-Nummer: 86123-10-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...