Ethyl-3-methyl-3-phenylglycidat (Gemisch der cis- und trans-Isomeren), Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Ethyl 3-Methyl-3-phenylglycidate (mixture of isomers), Ethylmethylphenylglycidate Ethyl 3-methyl-3-phenylglycidate 3-Methyl-3-phenyl glycidic acid ethyl ester, Ethyl 2,3-Epoxy-3-phenylbutyrate (mixture of isomers), 2,3-Epoxy-3-phenylbutyric acid Ethyl Ester (mixture of isomers), 3-Methyl-3-phenylglycidic acid Ethyl Ester (mixture of isomers)
M-Clarity™ quality level = MQ300
Formel:
C₁₂H₁₄O₃ |
MDL Number:
MFCD00022338 CAS-Nummer: 77-83-8 EINECS: 201-061-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...