Hexaamminruthenium(II)chlorid 99.9%

Lieferant: Thermo Fisher Scientific
Achtung

B23886.01 B23886.03
B23886.01EA 68.3 EUR
B23886.01 B23886.03
Hexaamminruthenium(II)chlorid 99.9%
Hexaammineruthenium(II) chloride

Purity: 99.9% CAS No.: 15305-72-3 Molecular Formula: Ru(NH3)6Cl2 Formula Weight: 274.16 Storage Temperature: 2°C to 8°C Odour: Odourless MDL No.: MFCD00011477

Formel: H₁₈Cl₂N₆Ru
Schmelzpunkt: >300 °C
MDL Number: MFCD00011477
CAS-Nummer: 15305-72-3
EINECS: 239-342-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR