Bis(2,2,6,6-tetramethyl-3,5-heptandionato-O,O')kupfer(II) ≥99,9% (Metall-Basis)
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Cu(TMHD)2, Copper(II)bis(2,2,6,6-tetramethyl-3,5-heptanedionate), Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)copper(II)
41279.03
41279.06
41279.03EA
88.6
EUR
41279.03
41279.06
Bis(2,2,6,6-tetramethyl-3,5-heptandionato-O,O')kupfer(II) ≥99,9% (Metall-Basis)
Bis(2,2,6,6-tetramethyl-3,5-heptandionato-O,O')kupfer(II)
Purity: 99.9%
CAS No.: 14040-05-2
Molecular Formula: C22H38CuO4
Formula Weight: 430.05
Physical Form: Powder
Melting Point: 198°
Boiling Point: 100°/0.1mm subl.
MDL No.: MFCD00058920
Formel:
C₂₂H₃₈CuO₄ Molekulargewicht: 430.08 g/mol Siedepunkt: 100 °C (0.1 mmHg) (Subl.) Schmelzpunkt: 198 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00058920 CAS-Nummer: 14040-05-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...