Encode syringe tips for repetitive pipettes, electronic/manual, AutoRep™
Lieferant: RAININ
ENC-2500S ENC-50ML ENC-5ML ENC-2500 ENC-25ML ENC-500 ENC-100 ENC-1250 ENC-12ML 17001871 17001873 17001883 17001879 17007399 17001881 17001877 ENC-1250S ENC-25MLS ENC-12MLS ENC-SET ENC-5MLS ENC-50MLS ENC-100S 17007400 17001874 17001875 17001876 17001880 17001872 17001884
RAINENC-2500SEA 521 EUR
RAINENC-2500S RAINENC-50ML RAINENC-5ML RAINENC-2500 RAINENC-25ML RAINENC-500 RAINENC-100 RAINENC-1250 RAINENC-12ML RAIN17001871 RAIN17001873 RAIN17001883 RAIN17001879 RAIN17007399 RAIN17001881 RAIN17001877 RAINENC-1250S RAINENC-25MLS RAINENC-12MLS RAINENC-SET RAINENC-5MLS RAINENC-50MLS RAINENC-100S RAIN17007400 RAIN17001874 RAIN17001875 RAIN17001876 RAIN17001880 RAIN17001872 RAIN17001884
Encode syringe tips for repetitive pipettes, electronic/manual, AutoRep™
Pipettenspitzen
These syringe tips are designed for use with AutoRep E and AutoRep S type manual and electronic pipettes. 25 and 50 ml syringe tips include an adapter.
  • Ideal for viscous or high vapor pressure liquids
  • No volume setting errors - syringe is automatically recognized by AutoRep E
  • Positive displacement: All liquid samples can be used, aqueous and non-aqueous
  • Sterile syringe tips are endotoxin-free and individually wrapped