96-Well micro test plates
Lieferant: SARSTEDT
82.1581 82.1581.001 82.1582 82.1582.001 82.1583 82.1583.001
SARS82.1581EA 85.8 EUR
SARS82.1581 SARS82.1581.001 SARS82.1582 SARS82.1582.001 SARS82.1583 SARS82.1583.001
96-Well micro test plates
Mikrotiterplatten HTS-Mikrotiterplatten
Clear PS
  • Compatible with all common microplate dispensers, readers, and washers
  • Excellent and consistent optical quality
  • Optimal surface
  • Uniform design
  • Choice of flat, round or conical wells