3-(1,1-Difluoroethyl)benzoic acid 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC57526-250MG
PC57526-1G
PC57526-5G
APOSPC57526-250MGEA
190
EUR
APOSPC57526-250MG
APOSPC57526-1G
APOSPC57526-5G
3-(1,1-Difluoroethyl)benzoic acid 95%
3-(1,1-Difluoroethyl)benzoic acid
Formel:
C₉H₈F₂O₂ |
MDL Number:
MFCD20640021 CAS-Nummer: 55805-17-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...