2,3-Difluoro-1,4-benzenediol 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2,3-Difluorohydroquinone
PC408062-250MG
PC408062-1G
APOSPC408062-250MGEA
52.2
EUR
APOSPC408062-250MG
APOSPC408062-1G
2,3-Difluoro-1,4-benzenediol 97%
2,3-Difluoro-1,4-benzenediol
Formel:
C₆H₄F₂O₂ Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD04038791 CAS-Nummer: 124728-90-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...