Sulpho-Cyanine 5 carboxylic acid ≥95%
Lieferant: AAT BIOQUEST
266
267
AATB266EA
769
EUR
AATB266
AATB267
Sulpho-Cyanine 5 carboxylic acid ≥95%
Sulpho-Cyanine 5 carboxylic acid
A variety of cyanine dyes has been used to label biological molecules for fluorescence imaging and other fluorescence-based biochemical analysis.
- pH Tolerance From 3-10
- Bright Fluorescence
- Multiple Bioconjugation Applications
Bitte beachten For research use only.
Formel:
C32H37N2NaO8S2 |
CAS-Nummer:
1121756-16-8 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...