2-(Trifluormethyl)benzoesäurehydrazid

Lieferant: Thermo Scientific

Synonyme: 2-(Trifluoromethyl)benzohydrazide, 2-(Trifluoromethyl)benzhydrazide

TL00781.1 TL00781.10 TL00781.50
MAYBTL00781.1EA 0 EUR
MAYBTL00781.1 MAYBTL00781.10 MAYBTL00781.50
2-(Trifluormethyl)benzoesäurehydrazid
2-(Trifluoromethyl)benzoic acid hydrazide
Formel: C₈H₇F₃N₂O
MDL Number: MFCD00221473
CAS-Nummer: 344-95-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR